siRNA Details
| Gene Symbol: Mrps24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026080.2 | NP_080356.1 | 4 | 466 | ||
| GenBank mRNA | AK003117.1 | - | 4 | 450 | ||
| AK007338.1 | - | 4 | 450 | |||
| AK010590.1 | - | 4 | 447 | |||
| AK012461.1 | - | 4 | 455 | |||
| AK014099.1 | - | 4 | 453 | |||
| AK215321.1 | - | 1 | 93 | |||
| BC029262.1 | - | 4 | 440 | 4911699 | ||

Mrps24
Genomic Map