siRNA Details
| Gene Symbol: Krtdap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033131.3 | NP_001028303.1 | 3 | 183 | ||
| GenBank mRNA | AK131603.1 | - | 3 | 184 | ||
| AK131606.1 | - | 3 | 184 | |||
| BC132510.1 | - | 3 | 183 | 40130787 | ||
| BC132512.1 | - | 3 | 183 | 40130789 | ||
| BC145438.1 | - | 3 | 183 | 9053996 | ||

Krtdap
Genomic Map