siRNA Details
| Gene Symbol: Lpin2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164885.1 | NP_001158357.1 | 11 | 1812 | ||
| NM_022882.4 | NP_075020.2 | 11 | 1688 | |||
| GenBank mRNA | AF286723.1 | - | 11 | 1688 | ||
| AK048657.1 | - | 1 | 1683 | |||
| BC039698.1 | - | 11 | 1801 | 5101211 | ||

Lpin2
Genomic Map