siRNA Details
| Gene Symbol: Scel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022886.2 | NP_075024.2 | 22 | 1421 | ||
| GenBank mRNA | AF245700.2 | - | 22 | 1421 | ||
| BC111874.1 | - | 22 | 1341 | 40059586 | ||
| BC112380.1 | - | 21 | 1282 | 40059585 | ||

Scel
Genomic Map