siRNA Details
| Gene Symbol: Izumo1r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022888.2 | NP_075026.1 | 3 | 455 | ||
| NM_176807.4 | NP_789777.1 | 3 | 455 | |||
| GenBank mRNA | AF250145.1 | - | 3 | 455 | ||
| AK003087.1 | - | 2 | 262 | |||
| AK041596.1 | - | 3 | 389 | |||
| AK083306.1 | - | 3 | 456 | |||
| BC028431.1 | - | 3 | 436 | 1265235 | ||
| EU326437.1 | - | 2 | 213 | |||
| EU326438.1 | - | 2 | 213 | |||

Izumo1r
Genomic Map