siRNA Details
| Gene Symbol: Pes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022889.3 | NP_075027.1 | 11 | 1169 | ||
| GenBank mRNA | AF289539.1 | - | 11 | 1129 | ||
| AK029075.1 | - | 11 | 1169 | |||
| AK088831.1 | - | 11 | 1147 | |||
| BC004844.1 | - | 11 | 1154 | 2649598 | ||
| BC011142.1 | - | 11 | 1152 | 4207164 | ||

Pes1
Genomic Map