siRNA Details
| Gene Symbol: Irak1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022986.4 | NP_075362.3 | 2 | 566 | ||
| GenBank mRNA | AF093135.1 | - | 2 | 349 | ||
| AJ242722.1 | - | 2 | 191 | |||
| AK013036.2 | - | 2 | 371 | |||
| AK020170.1 | - | 2 | 569 | |||
| AK020448.1 | - | 2 | 341 | |||
| AK045436.1 | - | 2 | 362 | |||
| AK076594.1 | - | 2 | 374 | |||
| BC046301.1 | - | 2 | 360 | |||
| BC061099.1 | - | 2 | 353 | 6742251 | ||

Irak1bp1
Genomic Map