siRNA Details
| Gene Symbol: Zic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022987.3 | NP_075363.1 | 2 | 1873 | ||
| GenBank mRNA | AB042155.1 | - | 2 | 1895 | ||
| AB114214.1 | - | 2 | 1532 | |||
| AK006747.1 | - | 2 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zic5| Gene Symbol: Zic5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022987.3 | NP_075363.1 | 2 | 1873 | ||
| GenBank mRNA | AB042155.1 | - | 2 | 1895 | ||
| AB114214.1 | - | 2 | 1532 | |||
| AK006747.1 | - | 2 | 271 | |||