siRNA Details
| Gene Symbol: Lrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022993.3 | NP_075369.2 | 5 | 1360 | ||
| GenBank mRNA | AB042200.1 | - | 5 | 1262 | ||
| AK132169.1 | - | 1344 | ||||
| AK144035.1 | - | 5 | 1373 | |||
| AK170325.1 | - | 5 | 1334 | |||
| AK194960.1 | - | 1 | 143 | |||
| AK203471.1 | - | 1 | 152 | |||
| BC052378.1 | - | 5 | 1336 | 5696275 | ||
| BC054447.1 | - | 5 | 1328 | 6477698 | ||

Lrp10
Genomic Map