siRNA Details
| Gene Symbol: Lrp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022993.3 | NP_075369.2 | 5 | 866 | ||
| GenBank mRNA | AB042200.1 | - | 5 | 768 | ||
| AK132169.1 | - | 850 | ||||
| AK144035.1 | - | 5 | 879 | |||
| AK170325.1 | - | 5 | 840 | |||
| AK185029.1 | - | 3 | 332 | |||
| AK185527.1 | - | 3 | 295 | |||
| AK197457.1 | - | 3 | 306 | |||
| AK203282.1 | - | 3 | 295 | |||
| BC052378.1 | - | 5 | 842 | 5696275 | ||
| BC054447.1 | - | 5 | 834 | 6477698 | ||

Lrp10
Genomic Map