siRNA Details
| Gene Symbol: Vps35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022997.4 | NP_075373.1 | 6 | 642 | ||
| GenBank mRNA | AF226323.1 | - | 6 | 661 | ||
| AK135680.1 | - | 6 | 948 | |||
| AK142382.1 | - | 6 | 929 | |||
| AK145421.1 | - | 5 | 609 | |||
| AK145853.1 | - | 6 | 691 | |||
| AK150682.1 | - | 6 | 656 | |||
| AK162726.1 | - | 6 | 621 | |||
| AK166821.1 | - | 6 | 635 | |||
| AK167421.1 | - | 6 | 659 | |||
| AK167594.1 | - | 6 | 655 | |||
| AK168880.1 | - | 6 | 635 | |||
| AK170473.1 | - | 6 | 676 | |||
| BC005469.1 | - | 6 | 667 | 3492842 | ||
| BC006637.1 | - | 6 | 647 | 3969440 | ||
| U47024.1 | - | 5 | 607 | |||

Vps35
Genomic Map