siRNA Details
Gene Symbol: Vps35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022997.4 | NP_075373.1 | 7 | 795 | ||
GenBank mRNA | AF226323.1 | - | 7 | 814 | ||
AK135680.1 | - | 7 | 1101 | |||
AK142382.1 | - | 7 | 1082 | |||
AK145421.1 | - | 6 | 762 | |||
AK145853.1 | - | 7 | 844 | |||
AK150682.1 | - | 7 | 809 | |||
AK162726.1 | - | 7 | 774 | |||
AK166821.1 | - | 7 | 788 | |||
AK167421.1 | - | 7 | 812 | |||
AK167594.1 | - | 7 | 808 | |||
AK168880.1 | - | 7 | 788 | |||
AK170473.1 | - | 7 | 829 | |||
AK218937.1 | - | 1 | 47 | |||
BC005469.1 | - | 7 | 820 | 3492842 | ||
BC006637.1 | - | 7 | 800 | 3969440 | ||
U47024.1 | - | 6 | 763 |
