siRNA Details
| Gene Symbol: Slc15a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023044.2 | NP_075531.2 | 5 | 1618 | ||
| GenBank mRNA | AF121080.1 | - | 5 | 1626 | ||
| AK051941.1 | - | 5 | 1219 | |||
| AK171698.1 | - | 1271 | ||||
| BC051940.1 | - | 5 | 1289 | 5717926 | ||

Slc15a3
Genomic Map