siRNA Details
| Gene Symbol: Asb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023048.5 | NP_075535.1 | 3, 4 | 526 | ||
| GenBank mRNA | AK018373.1 | - | 2, 3 | 679 | ||
| AK076004.1 | - | 2, 3 | 583 | |||
| AK133400.1 | - | 1, 2 | 129 | |||
| AK133894.1 | - | 2, 3 | 397 | |||
| BC046819.1 | - | 3, 4 | 526 | 5703017 | ||
| BC054745.1 | - | 2, 3 | 634 | 6836490 | ||
| BC056440.1 | - | 3, 4 | 526 | 5703017 | ||

Asb4
Genomic Map