siRNA Details
| Gene Symbol: Tmem176a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001098271.1 | NP_001091741.1 | 2 | 181 | ||
| NM_025326.4 | NP_079602.4 | 2 | 233 | |||
| GenBank mRNA | AB063313.1 | - | 1 | 123 | ||
| AK002546.1 | - | 2 | 209 | |||
| AK004315.1 | - | 2 | 206 | |||
| AK079378.1 | - | 195 | ||||
| AK150576.1 | - | 2 | 198 | |||
| AK162991.1 | - | 2 | 168 | |||
| AK165660.1 | - | 2 | 207 | |||
| BC010831.1 | - | 2 | 177 | 4221626 | ||
| BY352519.1 | - | 2 | 184 | |||
| CT010190.1 | - | 1 | 123 | |||

Tmem176a
Genomic Map