siRNA Details
| Gene Symbol: Chchd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025336.1 | NP_079612.1 | 1, 2 | 154 | ||
| GenBank mRNA | AK002263.1 | - | 1, 2 | 219 | ||
| AK002889.1 | - | 1, 2 | 226 | |||
| AK003519.1 | - | 1, 2 | 154 | |||
| AK006643.1 | - | 1, 2 | 220 | |||
| AK145337.1 | - | 1, 2 | 203 | |||
| BC021941.1 | - | 1, 2 | 206 | 5124504 | ||
| BC032987.1 | - | 1, 2 | 169 | |||

Chchd3
Genomic Map