siRNA Details
Gene Symbol: Ndufa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025348.2 | NP_079624.1 | 3 | 149 | ||
GenBank mRNA | AK003133.1 | - | 3 | 140 | ||
AK019099.1 | - | 3 | 151 | |||
BC061002.1 | - | 142 | ||||
BC120712.1 | - | 3 | 143 | 40129635 | ||
BC120714.1 | - | 3 | 143 | 40129637 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
