siRNA Details
| Gene Symbol: Ghitm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199122.1 | NP_001186051.1 | 8 | 1142 | ||
| NM_078478.5 | NP_510963.1 | 8 | 1095 | |||
| GenBank mRNA | AF412297.1 | - | 8 | 1039 | ||
| AK005636.1 | - | 8 | 1040 | |||
| AK093887.1 | - | 8 | 1071 | |||
| AK134592.1 | - | 8 | 1025 | |||
| AK146433.1 | - | 8 | 1002 | |||
| AK146853.1 | - | 8 | 1058 | |||
| AK151488.1 | - | 8 | 1072 | |||
| AK158904.1 | - | 8 | 1085 | |||
| AK159695.1 | - | 8 | 1069 | |||
| AK159724.1 | - | 8 | 1069 | |||
| AK164122.1 | - | 8 | 1040 | |||
| AK167970.1 | - | 8 | 1066 | |||
| AK168022.1 | - | 8 | 1072 | |||
| AK168678.1 | - | 8 | 1143 | |||
| AK168800.1 | - | 8 | 1051 | |||
| AK210138.1 | - | 1 | 25 | |||
| BC008622.1 | - | 8 | 1073 | 3588758 | ||
| BC010224.1 | - | 8 | 1075 | 3500685 | ||

Ghitm
Genomic Map