siRNA Details
| Gene Symbol: Smpx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252591.2 | NP_001239520.1 | 5 | 689 | ||
| GenBank mRNA | AF364070.1 | - | 6 | 563 | ||
| AJ245772.1 | - | 6 | 579 | |||
| AK003105.1 | - | 6 | 571 | |||
| AK009857.1 | - | 5 | 530 | |||
| AK010172.1 | - | 6 | 571 | |||
| AY026524.1 | - | 6 | 570 | |||
| BC051197.1 | - | 6 | 523 | 1397202 | ||
| BC061051.1 | - | 6 | 485 | 30300606 | ||
| BC132358.1 | - | 6 | 399 | 40130635 | ||
| BC138801.1 | - | 6 | 399 | 8861823 | ||

Smpx
Genomic Map