siRNA Details
| Gene Symbol: Wfdc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183249.2 | NP_899072.1 | 3 | 404 | ||
| GenBank mRNA | AA254806.1 | - | 3 | 334 | ||
| AK003164.1 | - | 3 | 346 | |||
| AK003352.1 | - | 3 | 348 | |||
| BC096415.1 | - | 3 | 339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wfdc21| Gene Symbol: Wfdc21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183249.2 | NP_899072.1 | 3 | 404 | ||
| GenBank mRNA | AA254806.1 | - | 3 | 334 | ||
| AK003164.1 | - | 3 | 346 | |||
| AK003352.1 | - | 3 | 348 | |||
| BC096415.1 | - | 3 | 339 | |||