siRNA Details
| Gene Symbol: Ndufa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025358.3 | NP_079634.2 | 3 | 273 | ||
| GenBank mRNA | AK003137.1 | - | 3 | 255 | ||
| AK003214.1 | - | 3 | 246 | |||
| AK012366.1 | - | 3 | 253 | |||
| BC005760.1 | - | 2 | 154 | |||
| BC058378.1 | - | 3 | 241 | 6820787 | ||

Ndufa9
Genomic Map