siRNA Details
| Gene Symbol: Cml1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023160.2 | NP_075649.2 | 2 | 847 | ||
| GenBank mRNA | AF163314.1 | - | 2 | 845 | ||
| AK002975.1 | - | 1 | 201 | |||
| AK003301.1 | - | 2 | 797 | |||
| BC010796.1 | - | 2 | 801 | 4223199 | ||

Cml1
Genomic Map