siRNA Details
| Gene Symbol: 1110001J03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025363.3 | NP_079639.1 | 1 | 120 | ||
| GenBank mRNA | AK003231.2 | - | 1 | 141 | ||
| AK008291.2 | - | 1 | 121 | |||
| AK009186.1 | - | 1 | 114 | |||
| BC059710.1 | - | 1 | 136 | 6539235 | ||

1110001J03Rik
Genomic Map