siRNA Details
| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025364.2 | NP_079640.1 | 9 | 639 | ||
| GenBank mRNA | AK003453.1 | - | 9 | 491 | ||
| AK018773.1 | - | 9 | 502 | |||
| AK088266.1 | - | 9 | 488 | |||
| AK146248.1 | - | 9 | 517 | |||
| BC027510.1 | - | 9 | 478 | 1382354 | ||
| BC094393.1 | - | 10 | 670 | |||
| BC157909.1 | - | 9 | 503 | 9088111 | ||
| BC158023.1 | - | 9 | 503 | 9007329 | ||
| BC172063.1 | - | 9 | 500 | 9054349 | ||

Sarnp
Genomic Map