siRNA Details
| Gene Symbol: Sarnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025364.2 | NP_079640.1 | 8, 9 | 605 | ||
| GenBank mRNA | AK003453.1 | - | 8, 9 | 457 | ||
| AK018773.1 | - | 8, 9 | 468 | |||
| AK088266.1 | - | 8, 9 | 454 | |||
| AK146248.1 | - | 8, 9 | 483 | |||
| BC027510.1 | - | 8, 9 | 444 | 1382354 | ||
| BC157909.1 | - | 8, 9 | 469 | 9088111 | ||
| BC158023.1 | - | 8, 9 | 469 | 9007329 | ||
| BC172063.1 | - | 8, 9 | 466 | 9054349 | ||

Sarnp
Genomic Map