siRNA Details
| Gene Symbol: Tomm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164729.1 | NP_001158201.1 | 3 | 634 | ||
| GenBank mRNA | BC021590.1 | - | 2 | 764 | 5068609 | |
| BC089325.1 | - | 1 | 1428 | |||
| CA319049.1 | - | 3 | 283 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tomm6| Gene Symbol: Tomm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164729.1 | NP_001158201.1 | 3 | 634 | ||
| GenBank mRNA | BC021590.1 | - | 2 | 764 | 5068609 | |
| BC089325.1 | - | 1 | 1428 | |||
| CA319049.1 | - | 3 | 283 | |||