siRNA Details
| Gene Symbol: Chchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025366.3 | NP_079642.1 | 1 | 129 | ||
| GenBank mRNA | AK003263.1 | - | 1 | 88 | ||
| AK010311.1 | - | 1 | 72 | |||
| AK011261.1 | - | 1 | 87 | |||
| AK027907.1 | - | 1 | 99 | |||
| AV460497.1 | - | 1 | 129 | |||
| BC049582.1 | - | 98 | 6772118 | |||
| BC052720.1 | - | 1 | 108 | 6818266 | ||
| BC064310.1 | - | 1 | 108 | |||
| BC092216.1 | - | 1 | 96 | 30609530 | ||

Chchd1
Genomic Map