siRNA Details
| Gene Symbol: Sf3b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175102.4 | NP_780311.2 | 1 | 374 | ||
| GenBank mRNA | AK003475.1 | - | 1 | 343 | ||
| AK009874.1 | - | 1 | 337 | |||
| AK010348.1 | - | 1 | 349 | |||
| BB652104.1 | - | 1 | 374 | |||
| BC006603.1 | - | 1 | 370 | 3965951 | ||

Sf3b5
Genomic Map