siRNA Details
| Gene Symbol: Aaed1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025370.2 | NP_079646.1 | 3 | 326 | ||
| GenBank mRNA | AK003782.1 | - | 3 | 327 | ||
| AK006589.1 | - | 3 | 181 | |||
| AK145073.1 | - | 324 | ||||
| AK145867.1 | - | 327 | ||||
| AK148379.1 | - | 327 | ||||
| AK168701.1 | - | 205 | ||||

Aaed1
Genomic Map