siRNA Details
| Gene Symbol: Tmem8c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025376.3 | NP_079652.1 | 1 | 216 | ||
| GenBank mRNA | AK003298.1 | - | 1 | 218 | ||
| AK004477.1 | - | 1 | 185 | |||
| BB839715.1 | - | 1 | 218 | |||
| BC062145.1 | - | 1 | 142 | 30286808 | ||

Tmem8c
Genomic Map