siRNA Details
| Gene Symbol: Ifitm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025378.2 | NP_079654.1 | 2 | 374 | ||
| GenBank mRNA | AK003407.1 | - | 2 | 375 | ||
| AK007916.1 | - | 2 | 367 | |||
| AK007919.1 | - | 2 | 363 | |||
| AK151890.1 | - | 2 | 340 | |||
| AY082484.1 | - | 2 | 343 | |||
| AY594690.1 | - | 2 | 390 | |||
| BC010291.1 | - | 2 | 326 | 3152819 | ||

Ifitm3
Genomic Map