siRNA Details
| Gene Symbol: Uqcr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_197979.2 | NP_932096.1 | 2 | 404 | ||
| GenBank mRNA | AK002227.1 | - | 2 | 394 | ||
| AK003881.1 | - | 2 | 401 | |||
| AK012489.1 | - | 2 | 397 | |||
| AK168491.1 | - | 2 | 378 | |||
| AW985484.1 | - | 2 | 372 | |||
| BC024518.1 | - | 2 | 388 | 4984172 | ||
| BC087874.1 | - | 2 | 394 | 6545811 | ||
| BC100591.1 | - | 2 | 394 | 30119194 | ||
| BX522336.1 | - | 3 | 454 | |||
| CB599515.1 | - | 2 | 407 | |||

Uqcr10
Genomic Map