siRNA Details
| Gene Symbol: Ufc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025388.2 | NP_079664.1 | 3 | 288 | ||
| GenBank mRNA | AK003070.1 | - | 2 | 190 | ||
| AK003897.1 | - | 3 | 289 | |||
| AK011260.1 | - | 2 | 179 | |||
| BC021936.1 | - | 3 | 249 | 5124904 | ||
| BC055109.1 | - | 1 | 333 | |||

Ufc1
Genomic Map