siRNA Details
| Gene Symbol: Mrpl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023167.2 | NP_075656.2 | 8 | 913 | ||
| GenBank mRNA | AB041599.1 | - | 7 | 665 | ||
| AB049634.1 | - | 8 | 912 | |||
| AK002464.1 | - | 8 | 749 | |||
| AK003746.2 | - | 8 | 870 | |||
| BC023078.1 | - | 5 | 1355 | |||
| BC037064.1 | - | 8 | 746 | |||
| BC039983.1 | - | 8 | 881 | 4952757 | ||
| BC051685.1 | - | 8 | 726 | |||
| BC061095.1 | - | 8 | 729 | 6588836 | ||

Mrpl4
Genomic Map