siRNA Details
| Gene Symbol: Grina | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023168.3 | NP_075657.1 | 4 | 721 | ||
| GenBank mRNA | AF182040.1 | - | 4 | 704 | ||
| AK185141.1 | - | 1 | 22 | |||
| AK188174.1 | - | 1 | 22 | |||
| AK189399.1 | - | 3 | 345 | |||
| AK218437.1 | - | 1 | 22 | |||
| BC010802.1 | - | 4 | 754 | 4236429 | ||
| BC019157.1 | - | 4 | 742 | |||
| BC037667.1 | - | 4 | 721 | 5376054 | ||

Grina
Genomic Map