siRNA Details
| Gene Symbol: Chchd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025395.3 | NP_079671.1 | 2 | 202 | ||
| GenBank mRNA | AK003341.1 | - | 2 | 104 | ||
| AK003936.1 | - | 2 | 85 | |||
| AK013018.1 | - | 2 | 81 | |||
| BC094414.1 | - | 2 | 93 | 1515083 | ||
| BC096668.1 | - | 2 | 100 | 1530928 | ||
| BY335381.1 | - | 2 | 205 | |||

Chchd5
Genomic Map