siRNA Details
| Gene Symbol: Chchd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025395.3 | NP_079671.1 | 4 | 620 | ||
| GenBank mRNA | AK003341.1 | - | 4 | 522 | ||
| AK003936.1 | - | 4 | 502 | |||
| AK013018.1 | - | 4 | 499 | |||
| BC027303.1 | - | 2 | 179 | |||
| BC094414.1 | - | 4 | 511 | 1515083 | ||
| BC096668.1 | - | 4 | 518 | 1530928 | ||

Chchd5
Genomic Map