siRNA Details
| Gene Symbol: Med11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025397.2 | NP_079673.2 | 3 | 358 | ||
| GenBank mRNA | AK003984.1 | - | 3 | 336 | ||
| AK008153.1 | - | 3 | 376 | |||
| BC025057.1 | - | 3 | 358 | 4238297 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Med11| Gene Symbol: Med11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025397.2 | NP_079673.2 | 3 | 358 | ||
| GenBank mRNA | AK003984.1 | - | 3 | 336 | ||
| AK008153.1 | - | 3 | 376 | |||
| BC025057.1 | - | 3 | 358 | 4238297 | ||