siRNA Details
| Gene Symbol: Mustn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181390.3 | NP_852055.1 | 3 | 522 | ||
| GenBank mRNA | AJ277212.1 | - | 3 | 658 | ||
| AK003360.1 | - | 3 | 238 | |||
| AK003945.1 | - | 3 | 240 | |||
| BC116939.1 | - | 3 | 230 | 40126258 | ||
| BC116941.1 | - | 3 | 230 | 40126260 | ||

Mustn1
Genomic Map