siRNA Details
| Gene Symbol: Nat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025400.3 | NP_079676.1 | 2, 3 | 294 | ||
| GenBank mRNA | AK003948.1 | - | 2, 3 | 248 | ||
| AK009262.1 | - | 1, 2 | 24 | |||
| AK020083.1 | - | 2, 3 | 274 | |||
| AK082469.1 | - | 2, 3 | 234 | |||
| AK186542.1 | - | 2, 3 | 134 | |||

Nat9
Genomic Map