siRNA Details
| Gene Symbol: Ubl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025401.3 | NP_079677.1 | 3 | 1711 | ||
| GenBank mRNA | AK002730.1 | - | 3 | 220 | ||
| AK003992.1 | - | 3 | 227 | |||
| AK007726.1 | - | 3 | 192 | |||
| AK009854.1 | - | 3 | 227 | |||
| AK012803.1 | - | 3 | 209 | |||
| AK051149.1 | - | 3 | 1712 | |||
| AK154036.1 | - | 2 | 2303 | |||
| BC028498.1 | - | 3 | 181 | 1400865 | ||

Ubl5
Genomic Map