siRNA Details
| Gene Symbol: 1110037F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081183.1 | NP_001074652.1 | 10, 11 | 2752 | ||
| GenBank mRNA | AK034232.1 | - | 9, 10 | 2524 | ||
| AK052499.1 | - | 9, 10 | 2540 | |||
| AK076731.1 | - | 9, 10 | 2572 | |||
| AK122509.1 | - | 9, 10 | 2505 | |||
| AK155623.1 | - | 9, 10 | 2603 | |||
| AK170075.1 | - | 9, 10 | 2605 | |||
| BC033309.1 | - | 10, 11 | 2722 | 5024976 | ||

1110037F02Rik
Genomic Map