siRNA Details
| Gene Symbol: Acer3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025408.2 | NP_079684.2 | 2 | 271 | ||
| GenBank mRNA | AK004287.1 | - | 2 | 272 | ||
| AK011668.1 | - | 2 | 252 | |||
| AK017361.1 | - | 176 | ||||
| AK080977.1 | - | 2 | 249 | |||
| AK150629.1 | - | 2 | 232 | |||
| AK162403.1 | - | 2 | 287 | |||
| BC023924.1 | - | 2 | 275 | 5324078 | ||

Acer3
Genomic Map