siRNA Details
| Gene Symbol: Pithd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025411.4 | NP_079687.3 | 2 | 337 | ||
| GenBank mRNA | AF400670.1 | - | 2 | 300 | ||
| AK004208.1 | - | 2 | 338 | |||
| AK050261.1 | - | 2 | 310 | |||
| BC052695.1 | - | 2 | 237 | 5715038 | ||
| BY044314.1 | - | 2 | 338 | |||

Pithd1
Genomic Map