siRNA Details
| Gene Symbol: 1110059E24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025423.2 | NP_079699.1 | 3, 4 | 406 | ||
| GenBank mRNA | AK004308.1 | - | 3, 4 | 407 | ||
| AK016266.1 | - | 3, 4 | 553 | |||
| BC023385.1 | - | 3, 4 | 403 | 5253886 | ||

1110059E24Rik
Genomic Map