siRNA Details
| Gene Symbol: Ndufb9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023172.3 | NP_075661.1 | 2 | 172 | ||
| GenBank mRNA | AK002254.1 | - | 2 | 173 | ||
| AK003540.1 | - | 2 | 173 | |||
| AK004522.1 | - | 2 | 173 | |||
| BC031539.1 | - | 2 | 154 | 3979845 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001127294, FQ217579, FQ217728, FQ210477, FQ224000, and others.

Ndufb9
Genomic Map