siRNA Details
| Gene Symbol: Mrps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025440.2 | NP_079716.1 | 2 | 247 | ||
| GenBank mRNA | AB049949.1 | - | 2 | 305 | ||
| AK005206.1 | - | 2 | 248 | |||
| AK019243.1 | - | 2 | 193 | |||
| BC024656.1 | - | 2 | 213 | 3986314 | ||

Mrps16
Genomic Map