siRNA Details
| Gene Symbol: Hspbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024172.3 | NP_077134.1 | 8 | 1194 | ||
| GenBank mRNA | AF338351.1 | - | 7 | 1132 | ||
| AK013263.1 | - | 8 | 1185 | |||
| AK088469.1 | - | 8 | 1170 | |||
| AK160128.1 | - | 8 | 1190 | |||
| AK209519.1 | - | 2 | 67 | |||
| BC014758.1 | - | 8 | 1194 | 4014661 | ||

Hspbp1
Genomic Map