siRNA Details
| Gene Symbol: Alg5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025442.3 | NP_079718.1 | 4, 5 | 403 | ||
| GenBank mRNA | AK003818.1 | - | 4, 5 | 384 | ||
| AK005296.1 | - | 4, 5 | 404 | |||
| AK006206.1 | - | 4, 5 | 372 | |||
| AK030567.1 | - | 4, 5 | 374 | |||
| AK161187.1 | - | 382 | ||||
| BC027160.1 | - | 4, 5 | 363 | 4946430 | ||

Alg5
Genomic Map