siRNA Details
| Gene Symbol: Aig1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025446.3 | NP_079722.1 | 4, 5 | 595 | ||
| GenBank mRNA | AK005332.1 | - | 4, 5 | 623 | ||
| AK008210.1 | - | 4, 5 | 598 | |||
| AK162306.1 | - | 4, 5 | 614 | |||
| BC035305.1 | - | 4, 5 | 745 | 3980923 | ||
| EF410113.1 | - | 2, 3 | 480 | |||

Aig1
Genomic Map